Learn More
TARGETMOL CHEMICALS INC Iobenguane sulfate 25MG
SDP
Catalog No. 502876256
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.Iobenguane sulfate (MIBG sulfate) is a high-affinity substrate for cholera toxin that disrupts cellular mono(ADP-ribosylation). Radioiodinated Iobenguane sulfate is used clinically as a tumor-targeted radiopharmaceutical for diagnosing and treating adrenergic tumors. Purity 99.04%
Catalog No. 50-287-6256 Supplier TARGETMOL CHEMICALS INC Supplier No. T1558825MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes.Iobenguane sulfate (MIBG sulfate) is a high-affinity substrate for cholera toxin that disrupts cellular mono(ADP-ribosylation). Radioiodinated Iobenguane sulfate is used clinically as a tumor-targeted radiopharmaceutical for diagnosing and treating adrenergic tumors. Purity 99.04%