Learn More
Hach Company Iodate-Iodide Digital Titrator Cartridge, 0.3998 N
Supplier:  Hach Company 1496101
Encompass
Iodate-Iodide Standard Solution. For determination of sulfite by Digital Titrator iodometric titration. Digits = mg/L SO3 in 20 mL sample. Digits x 0.0005 = meq. Approximately 40-100 tests
Catalog No. 50-465-318