Learn More
Aqua Solutions IODINE 0.1 NORMAL 500ML
SDP
Catalog No. NC0136995
Encompass_Preferred
Iodine 0.100 Normal Aqueous Solution Prepared To Astm E200 (apha)
This item is not returnable. View return policy
Catalog No. NC0136995 Supplier Aqua Solutions Supplier No. 4750500ML
Iodine 0.100 Normal Aqueous Solution Prepared To Astm E200 (apha)