Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Iodine, 1.0N Standardized Solution in ethanol

Catalog No. AA44488K2
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA44488K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA44488K2 Supplier Thermo Scientific™ Supplier No. 44488K2
 Restricted Item
Add to Cart

Specifications

CAS 7553-56-2
Physical Form Liquid
Quantity 1 L
Molecular Formula I{2}
MDL Number MFCD00011355
UN Number UN2924
Formula Weight 253.81
Concentration or Composition (by Analyte or Components) 0.90-1.10M
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.