Learn More
Revvity Health Sciences Inc Iodine-125 Radionuclide, 10mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14)
Catalog No. 502809922
Encompass
Iodine-125 Radionuclide, 10mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14)
Catalog No. 50-280-9922 Supplier Revvity Health Sciences Inc Supplier No. NEZ033010MC
Iodine-125 Radionuclide, 10mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14)