Learn More
Revvity Health Sciences Inc Iodine-125 Radionuclide, 1mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14)
Catalog No. 502812341
Encompass
Iodine-125 Radionuclide, 1mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14)
Catalog No. 50-281-2341 Supplier Revvity Health Sciences Inc Supplier No. NEZ033001MC
Iodine-125 Radionuclide, 1mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14)