Learn More
Revvity Health Sciences Inc Iodine-125 Radionuclide, 2mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14), Concentration: >350mCi/mL
Catalog No. 502810728
Encompass
Iodine-125 Radionuclide, 2mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14), Concentration: >350mCi/mL
Catalog No. 50-281-0728 Supplier Revvity Health Sciences Inc Supplier No. NEZ033H002MC
Iodine-125 Radionuclide, 2mCi, Specific Activity: ~17Ci/mg, 0.1M NaOH (pH 12-14), Concentration: >350mCi/mL