Learn More
Revvity Health Sciences Inc Iodine-125 Radionuclide, 5mCi, Specific Activity: ~17Ci/mg, 10-5M NaOH (pH 8-11)
Catalog No. 502812324
Encompass
Iodine-125 Radionuclide, 5mCi, Specific Activity: ~17Ci/mg, 10-5M NaOH (pH 8-11)
Catalog No. 50-281-2324 Supplier Revvity Health Sciences Inc Supplier No. NEZ033A005MC
Iodine-125 Radionuclide, 5mCi, Specific Activity: ~17Ci/mg, 10-5M NaOH (pH 8-11)