Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK1008125 Shop All Avantor Products
Change view
Click to view available options
Glass; 125g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-1008-125 Glass; 125g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MK-1008-125 Supplier Avantor Supplier No. 100802

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7553-56-2
Physical Form Crystalline
Quantity 125 g
Molecular Formula I2
MDL Number MFCD00011355 MFCD00164163
Synonym iodine, diiodine, iodine crystals, iodine sublimed, tincture iodine, vistarin, eranol, iodio, iodine solution, iode
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILES II
IUPAC Name diiodine
Molecular Weight (g/mol) 253.81
PubChem CID 807
ChEBI CHEBI:17606
Formula Weight 253.81
Grade ACS
Packaging Glass
Chemical Name or Material Iodine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.