Learn More
Sigma Aldrich Fine Chemicals Biosciences Iodine ACS reagent 99.8 so500G
Catalog No. 502897797
Encompass
Iodine exhibits exceptionally high dielectric polarization in various solvents (dioxane isobutylene p-xylene and benzene).
Catalog No. 50-289-7797 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 207772500G
Iodine exhibits exceptionally high dielectric polarization in various solvents (dioxane isobutylene p-xylene and benzene).