Learn More
Sigma Aldrich Fine Chemicals Biosciences Iodine ACS reagent 99.8 so5G
Catalog No. 502897652
Encompass
Iodine exhibits exceptionally high dielectric polarization in various solvents (dioxane isobutylene p-xylene and benzene).
Catalog No. 50-289-7652 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2077725G
Iodine exhibits exceptionally high dielectric polarization in various solvents (dioxane isobutylene p-xylene and benzene).