Learn More
Sigma Aldrich Fine Chemicals Biosciences Iodine ACS reagent 998 so25KG
Catalog No. 502903725
Encompass
Iodine exhibits exceptionally high dielectric polarization in various solvents (dioxane isobutylene p-xylene and benzene).
Catalog No. 50-290-3725 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 2077722.5KG
Iodine exhibits exceptionally high dielectric polarization in various solvents (dioxane isobutylene p-xylene and benzene).