Learn More
Volu Sol Iodine Alcohol (Ethanol 70% Dilutant Included), 10 mL & 16 oz / 500 mL, Volu-Sol™
SDP
Catalog No. 50420829 Shop All Volu Sol Products
Encompass_Preferred
Iodine Alcohol, commonly used to remove the mercury deposits in Schaudinn or PVA fixed fecal smears.  Prevents mercury-caused artifacts on slides during the Wheatley Trichrome staining procedure.  Component of Volu-Sol's Parasitology Kit.  This is a two-part item
Catalog No. 50-420-829 Supplier Volu Sol Supplier No. VXI016
Iodine Alcohol, commonly used to remove the mercury deposits in Schaudinn or PVA fixed fecal smears.  Prevents mercury-caused artifacts on slides during the Wheatley Trichrome staining procedure.  Component of Volu-Sol's Parasitology Kit.  This is a two-part item