Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine-Bromine Solution, Hanus (AOAC) for Iodine Absorption Number of Fats and Oils, Reagents

Catalog No. 01336475
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-475 500 mL
01-336-476 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-475 Supplier Reagents Holdings Llc Supplier No. C7218000500C
Add to Cart
Add to Cart

Iodine-Bromine Solution, Hanus (AOAC) for Iodine Absorption Number of Fats and Oils, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Iodine-Bromine Solution, Hanus (AOAC) for Iodine Absorption Number of Fats and Oils
Boiling Point 118°C
Color Brown
CAS 64-19-7, 7553-56-2, 7553-56-2
pH <2
Physical Form Liquid
Quantity 500 mL
Grade Reagent
Packaging Glass Bottle
Specific Gravity 1.07
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.