Learn More
Sigma Aldrich Fine Chemicals Biosciences Iodine flakes ReagentPlusR25KG
Catalog No. 502907027
Encompass
Iodine is a micro nutrient essential for thyroid synthesis. It is a component of Grams iodine which has been reported to be used in identifying cellulase-producing microorganisms. Photogeneration of iodine from anatase TiO2 has been studied.
Catalog No. 50-290-7027 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 3765582.5KG
Iodine is a micro nutrient essential for thyroid synthesis. It is a component of Grams iodine which has been reported to be used in identifying cellulase-producing microorganisms. Photogeneration of iodine from anatase TiO2 has been studied.