Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine (Iodine-Iodide), 0.0315 N, Ricca Chemical
SDP

Catalog No. R39850004C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R39850004C 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R39850004C Supplier Ricca Chemical Company Supplier No. R39850004C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Iodine
Boiling Point 100°C
Melting Point 0°C
CAS 7732-18-5, 7681-11-0, 7553-56-2
CAS Min % 0.4, 1.25, 98.36
CAS Max % 0.4, 1.25, 98.36
Physical Form Liquid
Quantity 4 L
Assay Percent Range 0.0315 to 0.0315
Assay Percent Notes Normality Iodine
Grade Laboratory
Packaging Amber Glass Bottle
Appearance Dark Brown Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.