Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine (Iodine-Iodide), 0.315 N, Ricca Chemical
SDP

Catalog No. R40120001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R40120001C 1 L Amber Glass Bottle
R40120004C 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. R40120001C Supplier Ricca Chemical Company Supplier No. R40120001C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Specifications

CAS 7553-56-2, 7681-11-0, 7553-56-2
CAS Max % 85.57, 10.95, 3.48
Quantity 1 L
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Concentration or Composition (by Analyte or Components) 0.3148 to 0.3152 N
Packaging Amber Glass Bottle
Appearance Dark Brown Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.