Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Iodine lump, ultra dry, 99.999% (metals basis)

Catalog No. AA4485722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4485722 100 g
AA4485706 5 g
AA4485714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AA4485722 Supplier Thermo Scientific™ Supplier No. 4485722
Add to Cart
This item is not returnable. View return policy

Specifications

Melting Point 183-186° subl.
CAS 7553-56-2
Physical Form Solid
Quantity 100 g
Molecular Formula I{2}
MDL Number MFCD00011355
UN Number UN3495
Formula Weight 253.81
Percent Purity 99.999%
Density 4.930
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.