Learn More
Sigma Aldrich Fine Chemicals Biosciences Iodine monochloride reagen100G
Catalog No. 502897628
Encompass
Iodine monochloride is an interhalogen compound. It forms various complexes with ethyl isopropyl and t-butylbenzenes. Electrically conducting SOLUTIONon of ICl is obtained on disSOLUTIONon of ICl in polar solvents. ICl is a green oxidizing agent and participates in the following transformationsaldose hemiacetals to the corresponding aldose lactonesdiarylmethanols to the corresponding diarylmethanonesarylalkylmethanols to the corresponding arylalkylmethanonesdialkylmethanols to the corresponding dialkylmethanones
Catalog No. 50-289-7628 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 208221100G
Iodine monochloride is an interhalogen compound. It forms various complexes with ethyl isopropyl and t-butylbenzenes. Electrically conducting SOLUTIONon of ICl is obtained on disSOLUTIONon of ICl in polar solvents. ICl is a green oxidizing agent and participates in the following transformationsaldose hemiacetals to the corresponding aldose lactonesdiarylmethanols to the corresponding diarylmethanonesarylalkylmethanols to the corresponding arylalkylmethanonesdialkylmethanols to the corresponding dialkylmethanones