Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iodine Monochloride Solution

Catalog No. 714836100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-483-6100ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-483-6100ML Supplier Sigma Aldrich Supplier No. 714836100ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iodine monochloride is an inorganic reagent used in the iodination and chloroiodination of aromatic and unsaturated compounds, respectively. It is also used to cleave carbon-metal bonds.Iodine monochloride can be used:As a reagent in the preparation of +a-iodo +a-ketosulfones from +a-ketosulfones.In the synthesis of 3-(4-bromo-2-methylphenyl)-4-iodosydnone, which is further used to prepare substituted pyrazoles.

Specifications

CAS 884879-24-7
Melting Point 127°C to 130°C
Molecular Formula C36H47ClN2Pd
Linear Formula C36H47ClN2Pd
Quantity 1 g
MDL Number MFCD09265339
Synonym CX 32; [1,3-Bis[2,6-bis(1-methylethyl)phenyl]-2-imidazolidinylidene]chloro[(1,2,3-eta)-3-phenyl-2-propenyl]palladium
Molecular Weight (g/mol) 649.64 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.