Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hardy Diagnostics™ Iodine, Non-stabilized
SDP

Catalog No. 23001792 Shop All Hardy Diagnostics Products
Change view
Click to view available options
Boiling Point:
184°C
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Boiling Point
23-001-792 184°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-001-792 Supplier Hardy Diagnostics™ Supplier No. I008N
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A mordant used in the gram stain procedure. A non-stabilized, thinner solution

  • Non-stabilized Iodine has a shorter shelf life, is thinner and decolorizes more quickly than Stabilized Iodine

Specifications

Format Liquid
Boiling Point 184°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.