Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Solution, SPEX CertiPrep™
SDP

Catalog No. 183134 Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
1 gal.
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-313-4 1 gal. Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-313-4 Supplier SPEX CertiPrep Supplier No. 183134

missing translation for 'validMainframeMsg'

Guaranteed clean and stable, the matrix is prepared using double-deionized, ASTM Type I Water. SPEX CertiPrep™ Iodine solutions are prepared from ACS Grade potassium iodide and crystalline elemental iodine.

  • Ideal for ASTM D1510 applications
  • Packaged in an amber glass bottle

Chemical Identifiers

CAS 7681-11-0 , 7732-18-5
Molecular Formula IK
Molecular Weight (g/mol) 166.003
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Physical Form Liquid
Quantity 1 gal.
Assay Percent Range Iodine: 99.8%, Potassium Iodide: 99%
Formula Weight Iodine: 126.90447 u, Potassium Iodide: 166.0028 g/mol
Concentration or Composition (by Analyte or Components) 0.0473 N
Packaging Amber Glass Bottle
Chemical Name or Material H2O, KI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.