Learn More
Iodine Solution, Dilute Lugol’s (1% w/v), Reagents
Specifications
Specifications
| Chemical Name or Material | Iodine Solution, Dilute Lugol’s (1% w/v) |
| Boiling Point | 100°C |
| CAS | 7732-18-5, 7681-11-0, 7681-11-0 |
| Color | Dark Brown |
| Physical Form | Liquid |
| Quantity | 100 mL |
| Packaging | Glass Bottle |
| Specific Gravity | 1.1 |
| Grade | Reagent |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.