Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Iodine Solution, Dilute Lugol’s (1% w/v), Reagents
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Chemical Name or Material | Iodine Solution, Dilute Lugol’s (1% w/v) |
Boiling Point | 100°C |
CAS | 7732-18-5, 7681-11-0, 7681-11-0 |
Color | Dark Brown |
Physical Form | Liquid |
Quantity | 100 mL |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction