Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodine Solution - Dilute Lugol's Solution, 1.8 %, Fisher Science Education
SCI_ED

Catalog No. S96870 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
10 mL
Packaging:
.05m Solution
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S96870 10 mL .05m Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96870

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7732-18-5

Specifications

Color Amber
Boiling Point 100
Packaging .05m Solution
Quantity 10 mL
Chemical Name or Material Iodine Solution
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.