Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hanus Solution, Fisher Chemical

Catalog No. SI98500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SI98-500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SI98-500

missing translation for 'validMainframeMsg'

Used for iodine number determination

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by an SFI certified supplier.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-33-5 , 64-19-7
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00011353
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym Iodine monobromide, Iodine bromide, Iodobromine, Iodine bromide, Iodine Solution
PubChem CID 24408
ChEBI CHEBI:29224
IUPAC Name bromide
SMILES [Br-]

Specifications

Color Brown
Boiling Point 118°C
Melting Point 17°C
Concentration 0.210 to 0.230
Density 1.06g/cm³
Identification Pass test
Packaging Glass Bottle
Vapor Pressure 11mmHg
pH 2.0
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Iodine Solution, Hanus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.