Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Iodine spheres, ultra dry, -10 mesh, 99.998% (metals basis)

Catalog No. AA1431214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1431214 25 g
AA1431206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1431214 Supplier Thermo Scientific™ Supplier No. 1431214
Add to Cart

Specifications

Melting Point 183-186° subl.
CAS 7553-56-2
Physical Form Solid
Quantity 25 g
Molecular Formula I{2}
MDL Number MFCD00011355
UN Number UN3495
Formula Weight 253.81
Percent Purity 99.998%
Density 4.930
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.