Learn More
Medchemexpress LLC Iodo-PEG3-N3 | 936917-36-1 | 97.0% | 100 MG
SDP
Catalog No. 5000339971
Encompass_Preferred
Iodo-PEG3-N3 is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs employ the intracellular ubiquitin-proteasome system to specifically degrade target proteins.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Enables specific degradation of target proteins
Catalog No. 50-003-39971 Supplier Medchemexpress LLC Supplier No. HY130525100MG
Iodo-PEG3-N3 is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent, containing an azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs employ the intracellular ubiquitin-proteasome system to specifically degrade target proteins.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Click chemistry reagent
  • Contains an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Enables specific degradation of target proteins