Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iodo-PEG7-alcohol | >98.0% | C14H29IO7 | 100 MG
SDP

Catalog No. 5000354930
Encompass_Preferred

Iodo-PEG7-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight of 436.28
  • Appears as a colorless to light yellow liquid
  • Store at -20°C, protected from light

Catalog No. 50-003-54930 Supplier Medchemexpress LLC Supplier No. HY143833100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iodo-PEG7-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one is a ligand for an E3 ubiquitin ligase and the other is for the target protein. PROTACs exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker
  • Can be used in the synthesis of PROTACs
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
  • Molecular weight of 436.28
  • Appears as a colorless to light yellow liquid
  • Store at -20°C, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.