Learn More
Medchemexpress LLC Iodo-PEG7-alcohol | 98.0% | C14H29IO7 | 50 MG
SDP
Catalog No. 5000355595
Encompass_Preferred
Iodo-PEG7-alcohol is a PEG-based PROTAC linker utilized in the synthesis of Proteolysis-Targeting Chimeras (PROTACs). PROTACs are designed to harness the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand is for an E3 ubiquitin ligase
  • The other ligand is for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-55595 Supplier Medchemexpress LLC Supplier No. HY14383350MG
Iodo-PEG7-alcohol is a PEG-based PROTAC linker utilized in the synthesis of Proteolysis-Targeting Chimeras (PROTACs). PROTACs are designed to harness the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Used in the synthesis of PROTACs
  • PROTACs contain two different ligands connected by a linker
  • One ligand is for an E3 ubiquitin ligase
  • The other ligand is for the target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins