Learn More
Abcam 5-(Iodoacetamido)fluorescein, 25MG
Catalog No. 50996130
Encompass_Preferred
MW 515.3 g/mol, Purity >90%. Specifically and efficiently labels thiol groups on proteins. Fluorescein derivative sucessfully links to organelles, proteins and peptides at or near their physiological pH values (pH 7), including enzymes (e.g. (Na, K)-ATPase) or organelles like nucleii and nuclear matrices.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-130 Supplier Abcam Supplier No. AB27389525MG
MW 515.3 g/mol, Purity >90%. Specifically and efficiently labels thiol groups on proteins. Fluorescein derivative sucessfully links to organelles, proteins and peptides at or near their physiological pH values (pH 7), including enzymes (e.g. (Na, K)-ATPase) or organelles like nucleii and nuclear matrices.
The product is subject to the following: Abcam Restricted Use Statement