Learn More
Medchemexpress LLC Iodoacetamide-PEG3-azide | 1594986-04-5 | 98.41% | C10H19IN4O4 | 250 MG
SDP
Catalog No. 5000352891
Encompass_Preferred
Iodoacetamide-PEG3-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Features an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-52891 Supplier Medchemexpress LLC Supplier No. 250MGHY140857
Iodoacetamide-PEG3-azide is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent features an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with Alkyne-containing molecules. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Features an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions