Learn More
Crescent Chemical Co Inc IODOACETAMIDE RESEARCH GRADE
SDP
Catalog No. 50247759
Encompass_Preferred
Iodoacetamide research grade Alkylating agent for use in protein sample preparation applications. For carboxymethylation of proteins. Size - 25G Storage Conditions - +2 °C TO +8 °C Catalog Number - 26710.02 CAS 144-48-9
Catalog No. 50-247-759 Supplier Crescent Chemical Co Inc Supplier No. SE26710.02
Iodoacetamide research grade Alkylating agent for use in protein sample preparation applications. For carboxymethylation of proteins. Size - 25G Storage Conditions - +2 °C TO +8 °C Catalog Number - 26710.02 CAS 144-48-9