Learn More
Crescent Chemical Co Inc IODOACETAMIDE RESEARCH GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE26710.01
Encompass_Preferred
Iodoacetamide research grade Alkylating agent for use in protein sample preparation applications. For carboxymethylation of proteins. Size - 5G Storage Conditions - +2 °C TO +8 °C Catalog Number - 26710.01 CAS 144-48-9
Catalog No. 50-247-758