Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT)

Catalog No. FERA52285 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 x 3.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERA52285 10 x 3.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERA52285 Supplier Thermo Scientific™ Supplier No. A52285
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Iodoacetamido-LC-Phosphonic Acid (6C-CysPAT) is a sulfhydryl-reactive alkylating reagent that contains a phosphonic acid group for enrichment of cysteine-containing peptides for mass spectrometry analysis.

  • A sulfhydryl-reactive alkylating reagent that contains a phosphonic acid group for enrichment of cysteine-containing peptides for mass spectrometry analysis
  • Provided in convenient single-use, no-weigh format
  • High-purity, crystalline reagent
  • Compatible with EasyPep chemistry and High Select Phosphopeptide Enrichment kits
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 x 3.5 mg
Molecular Weight (g/mol) 348.99
Percent Purity >90%
Grade MS grade
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.