Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iodoacetic Acid N-Hydroxysuccinimide Ester

Catalog No. I976050MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I976050MG 10 mg
I9760100MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. I976050MG Supplier Sigma Aldrich Supplier No. I976050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

A heterobifunctional cross-linking reagent with amine and sulfhydryl reactivity. Typically, coupled initially to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5) Second coupling specific for molecules containing free sulfydryl by thioether linkage buffered at pH 7.5 (7.0-8.0). Useful for preparation of enzyme immunoconjugates and hapten carrier molecule conjugates. Provides a 2-ato mLinker.

Specifications

Molecular Formula C10H15NO4
Linear Formula C10H15NO4
MDL Number MFCD00171363
Quantity 10 mg
Synonym N-(3-Oxohexanoyl)-L-homoserine lactone
Molecular Weight (g/mol) 213.23 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.