Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Iodoacetic anhydride | 54907-61-8 | MFCD00001080 | 70.0% | 353.88 | C4H4I2O3 | 25 MG
SDP

Catalog No. 5000284832
Encompass_Preferred

Iodoacetic anhydride is a halogenated anhydride reagent used in biochemical research for cysteine alkylation and protein modification. It is supplied as a powdered reagent for laboratory-scale chemical synthesis and protein derivatization.

  • Used for cysteine alkylation and protein modification.
  • Purity 70.0% (NMR).
  • Molecular weight 353.88.
  • Chemical formula C4H4I2O3.
  • CAS number 54907-61-8.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes suitable for lab use, including 25 MG, 50 MG, 100 MG.

Catalog No. 50-002-84832 Supplier Medchemexpress LLC Supplier No. HYW68660525MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Iodoacetic anhydride is a halogenated anhydride reagent used in biochemical research for cysteine alkylation and protein modification. It is supplied as a powdered reagent for laboratory-scale chemical synthesis and protein derivatization.

  • Used for cysteine alkylation and protein modification.
  • Purity 70.0% (NMR).
  • Molecular weight 353.88.
  • Chemical formula C4H4I2O3.
  • CAS number 54907-61-8.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in small pack sizes suitable for lab use, including 25 MG, 50 MG, 100 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.