Learn More
Apexbio Technology LLC Iodoacetyl-LC-Biotin,100mg CAS# 93285-75-7
SDP
Catalog No. 502240088
Encompass_Preferred
Iodoacetyl-LC-Biotin (N-iodoacetyl-N-biotinylhexylenediamine), a sulfhydryl-reactive biotinylation agent, is a water-insoluble reagent that requires the dissolution of suitable solvents, including dimethyl sulfoxide (DMSO) and dimethylformamide (DMF), prior to the addition into aqueous reactions. Iodoacetyl-LC-Biotin consists of a bicyclic biotin rings structure, a 1,6-diaminohexane spacer group attached to the valeric acid side chain of biotin and an iodoacetyl group at the end of the spacer providing sulfhydryl reactivity. The iodoacetyl group at the end of Iodoacetyl-LC-Biotin is able to react with sulfhydryl groups on proteins and other molecules forming a nonreversible bond. Other sizes are also available. Please inqury us for quote.
This item is not returnable. View return policy
Catalog No. 50-224-0088 Supplier Apexbio Technology LLC Supplier No. A8009100
Iodoacetyl-LC-Biotin (N-iodoacetyl-N-biotinylhexylenediamine), a sulfhydryl-reactive biotinylation agent, is a water-insoluble reagent that requires the dissolution of suitable solvents, including dimethyl sulfoxide (DMSO) and dimethylformamide (DMF), prior to the addition into aqueous reactions. Iodoacetyl-LC-Biotin consists of a bicyclic biotin rings structure, a 1,6-diaminohexane spacer group attached to the valeric acid side chain of biotin and an iodoacetyl group at the end of the spacer providing sulfhydryl reactivity. The iodoacetyl group at the end of Iodoacetyl-LC-Biotin is able to react with sulfhydryl groups on proteins and other molecules forming a nonreversible bond. Other sizes are also available. Please inqury us for quote.