Learn More
Medchemexpress LLC Iodobenzene | 591-50-4 | MFCD00001029 | 99.7% | 204.01 g/mol | C6H5I | 100g
SDP
Catalog No. 5000267924
Encompass_Preferred
Iodobenzene can undergo Ullmann coupling reaction on copper catalyst to generate biphenyl Iodobenzene can undergo Suzuki reaction with phenylboronic acid to generate biphenyl[1][2]
Catalog No. 50-002-67924 Supplier Medchemexpress LLC Supplier No. HYY0747100G
Iodobenzene can undergo Ullmann coupling reaction on copper catalyst to generate biphenyl Iodobenzene can undergo Suzuki reaction with phenylboronic acid to generate biphenyl[1][2]