Learn More
Medchemexpress LLC Iodobenzene | 591-50-4 | MFCD00001029 | 99.7% | 204.01 g/mol | C6H5I | 25g
SDP
Catalog No. 5000269819
Encompass_Preferred
Iodobenzene can undergo Ullmann coupling reaction on copper catalyst to generate biphenyl Iodobenzene can undergo Suzuki reaction with phenylboronic acid to generate biphenyl[1][2]
Catalog No. 50-002-69819 Supplier Medchemexpress LLC Supplier No. HYY074725G
Iodobenzene can undergo Ullmann coupling reaction on copper catalyst to generate biphenyl Iodobenzene can undergo Suzuki reaction with phenylboronic acid to generate biphenyl[1][2]