Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoprene, 98%, stabilized

Catalog No. AC122670025
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle

CAS: 78-79-5 | C5H8 | 68.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-79-5, 98-29-3
Molecular Formula C5H8
Molecular Weight (g/mol) 68.11
MDL Number MFCD00008600
InChI Key RRHGJUQNOFWUDK-UHFFFAOYSA-N
Synonym isoprene, 2-methyl-1,3-butadiene, isopentadiene, 2-methylbutadiene, 1,3-butadiene, 2-methyl, 2-methyldivinyl, beta-methylbivinyl, isopreno, isoterpene, isopren
PubChem CID 6557
ChEBI CHEBI:35194
IUPAC Name 2-methylbuta-1,3-diene
SMILES CC(=C)C=C

Specifications

Melting Point -146.0°C
Color Colorless to Yellow
Density 0.6800g/mL
Boiling Point 34.0°C
Flash Point −48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHC(CH3)=CH2
Refractive Index 1.4205 to 1.4225
Quantity 2.5 L
Beilstein 01, 252
Fieser 13,196
Specific Gravity 0.68
Merck Index 15, 5247
Solubility Information Solubility in water: 0.7g/L (25°C). Other solubilities: miscible with alcohol,ether and acetone,oils/fat
Stabilizer 100 to 250ppm 4-tert-butylcatechol
Viscosity 0.225 mPa.s (15°C)
Formula Weight 68.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Isoprene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause cancer.
Suspected of causing genetic defects.
Toxic to aquatic life with long lasting effects.
Extremely flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Obtain special instructions before use.
Wea
EINECSNumber 201-143-3
RTECSNumber NT4037000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.