Learn More
Medchemexpress LLC Iodobenzene-d5 | 7379-67-1 | 209.04 | C6D5I | 10g
SDP
Catalog No. 5000268597
Encompass_Preferred
Iodobenzene-d5 is the deuterium labeled 1-Iodobenzene Iodobenzene can undergo Ullmann coupling reaction on copper catalyst to generate biphenyl Iodobenzene can undergo Suzuki reaction with phenylboronic acid to generate biphenyl[1][2]
Catalog No. 50-002-68597 Supplier Medchemexpress LLC Supplier No. HYY0747S10G
Iodobenzene-d5 is the deuterium labeled 1-Iodobenzene Iodobenzene can undergo Ullmann coupling reaction on copper catalyst to generate biphenyl Iodobenzene can undergo Suzuki reaction with phenylboronic acid to generate biphenyl[1][2]