Learn More
American Radiolabeled Chemicals Inc IODOCYANOPIN 125I 100UC
SDP
Catalog No. 50901091
Encompass
Iodo-(-)-cyanopindolol-125I 100uCi in tris buffer, 0.1 mCi/ml, S.A. 2200 Ci/mmol, M.W. 411, Shipped in Dry Ice
Catalog No. 50-901-091 Supplier American Radiolabeled Chemicals Inc Supplier No. ARI 0186100UC
This product is regulated by the federal government, and may not be ordered online.
Iodo-(-)-cyanopindolol-125I 100uCi in tris buffer, 0.1 mCi/ml, S.A. 2200 Ci/mmol, M.W. 411, Shipped in Dry Ice