Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodomesitylene Diacetate 98.0+%, TCI America™
SDP

Catalog No. I04795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04795G Supplier TCI America Supplier No. I04795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33035-41-5
Molecular Formula C13H17IO4
MDL Number MFCD00674196
Synonym 2-(Diacetoxyiodo)mesitylene, 2,4,6-Trimethyl(diacetoxyiodo)benzene, 2,4,6-Trimethyliodobenzene Diacetate

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 5 g
Formula Weight 364.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Iodomesitylene Diacetate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.