Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iodomethane

Catalog No. I8507500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
10 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I8507500G 500 g
I8507100G 100 g
I850710KG 10 kg
I85072KG 2 kg
I85075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. I8507500G Supplier Sigma Aldrich Supplier No. I8507500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Iodomethane

Specifications

CAS 74-88-4
Density 2.28 g/mL at 25°C
Boiling Point 41°C to 43°C
Molecular Formula CH3I
Refractive Index n20/D 1.531
Linear Formula CH3I
MDL Number MFCD00001073
Quantity 500 g
Synonym Methyl iodide
Molecular Weight (g/mol) 141.94 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.