Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Iodomethane-d3, for NMR, 99+ atom % D

Catalog No. AC174950250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC174950250 25 g Glass Bottle
AC174950050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC174950250 Supplier Thermo Scientific Chemicals Supplier No. 174950250
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 865-50-9 | CH3I | 144.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 865-50-9
Molecular Formula CH3I
Molecular Weight (g/mol) 144.96
MDL Number MFCD00001074
InChI Key INQOMBQAUSQDDS-FIBGUPNXSA-N
Synonym iodomethane-d3, iodo 2h3 methane, trideuterio iodo methane, iodo 2 h? methane, methyl-d3 iodide, trideuteromethyl iodide, iodomethane-12c,d3, methyl-12c,d3 iodide, iodoethane-d5 ethyliodid-d5, methyl iodide d3 stored over copper
PubChem CID 2723978
IUPAC Name trideuterio(iodo)methane
SMILES [2H]C([2H])([2H])I

Specifications

Physical Form Liquid
Refractive Index 1.5262
Color Colorless
Quantity 25 g
Packaging Glass Bottle
Deuterium (D) 99.0% min.
Density 2.3290g/mL
Water 0.05% max. (Coulometric)
Boiling Point 42.0°C
Linear Formula CD3I
Formula Weight 144.96
Grade NMR
Chemical Name or Material Iodomethane-d3, 99+ atom % D
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Fatal if inhaled.
Harmful in contact with skin.
Suspected of causing cancer.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-744-5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.