Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Iodomethane Solution

Catalog No. 456756100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-675-6100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 45-675-6100ML Supplier Sigma Aldrich Supplier No. 456756100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product is 2M solution of iodomethane in tert-butyl methyl ether. Iodomethane also known as methyl iodide is an alkyl halide commonly employed as methylating agent.

Specifications

CAS 3587-60-8
Density 1.126 g/mL at 20°C
Boiling Point 102°C (14 mmHg, lit.)
Molecular Formula C8H9ClO
Linear Formula C6H5CH2OCH2Cl
Quantity 100 mL
MDL Number MFCD00000886
Synonym Methyl iodide
Molecular Weight (g/mol) 156.61 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.