Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Iodomethyl)cyclopropane, 95%

Catalog No. AAH3530903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3530903 1 g
AAH35309MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3530903 Supplier Thermo Scientific Chemicals Supplier No. H3530903
Only null left
Add to Cart

CAS: 33574-02-6 | C4H7I | 182.004 g/mol

(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Iodomethyl)cyclopropane is used as pharmaceutical intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33574-02-6
Molecular Formula C4H7I
Molecular Weight (g/mol) 182.004
MDL Number MFCD06410734
InChI Key JHKJQWFHNIOUKY-UHFFFAOYSA-N
Synonym iodomethyl cyclopropane, cyclopropylmethyl iodide, cyclopropane, iodomethyl, ambkkkkk549, acmc-20aplj, 2-iodomethyl cyclopropane, iodomethyl cyclopropane gc
PubChem CID 2763048
IUPAC Name iodomethylcyclopropane
SMILES C1CC1CI

Specifications

Quantity 1 g
Sensitivity Light sensitive
Formula Weight 182
Percent Purity 97%
Chemical Name or Material (Iodomethyl)cyclopropane
Hazard Category H319-H334
Hazard Statement GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P280i-P285-P304+P341-P305+P351+P338-P342+P311-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.