Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (Iodomethyl)Trimethylsilane

Catalog No. 5790325ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-903-25ML 25 mL
57-903-5ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 57-903-25ML Supplier Sigma Aldrich Supplier No. 5790325ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Starting material for the preparation of 2-(trimethylsilyl)ethyl-triphenylphosphonium iodide used for the synthesis of allyl silanes; Used for the N-alkylation of amides, in a reaction sequence to prepare unstabilized ylides for [2+3]-cycloadditions

Specifications

CAS 4206-67-1
Density 1.443 g/mL at 25°C
Boiling Point 139°C to 141°C
Molecular Formula C4H11ISi
Refractive Index n20/D 1.491; n20/D 1.491
Linear Formula (CH3)3SiCH2I
Quantity 25 mL
MDL Number MFCD00001077
Synonym (Trimethylsilyl)methyl iodide
Molecular Weight (g/mol) 214.12 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.