Learn More
Crescent Chemical Co Inc IODONITROTETRAZOLIUM CHLORIDE
SDP
Supplier:  Crescent Chemical Co Inc SE26840.03
Encompass_Preferred
Iodonitrotetrazolium chloride research grade For LDH detection (1).For colorimetric measurement of enzymatic hydrolysis of terminal galactose from GM1 ganglioside (2). Size - 10G Storage Conditions - +15 °C TO +30 °C Catalog Number - 26840.03 CAS 146-6
Catalog No. 50-247-761